4-(4-Fluorophenyl)benzonitrile 98%

Supplier: Ambeed
Danger

A803935-1G A803935-250MG A803935-5G
77006-604EA 75.99 USD
77006-604 77006-606 77006-608
4-(4-Fluorophenyl)benzonitrile 98%
4-(4-Fluorophenyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₃H₈FN
MDL Number: MFCD05981088
CAS Number: 10540-31-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR