N-Benzyl-3-bromoaniline 98%

Supplier: Ambeed
Warning

A803906-1G A803906-25G A803906-5G
77019-432EA 29.71 USD
77019-432 77019-434 77019-436
N-Benzyl-3-bromoaniline 98%
N-Benzyl-3-bromoaniline

Precaución: for Research Use Only

Formula: C₁₃H₁₂BrN
MW: 262.15 g/mol
MDL Number: MFCD11145017
CAS Number: 213814-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR