6-Amino-3-bromo-2-fluoropyridine 98%

Supplier: Ambeed
Warning

A803826-100MG A803826-1G A803826-250MG A803826-5G A803826-10G
76993-302EA 5 USD
76993-302 76993-304 76993-306 77552-470 77886-790
6-Amino-3-bromo-2-fluoropyridine 98%
6-Amino-3-bromo-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₄BrFN₂
MDL Number: MFCD11848480
CAS Number: 944401-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR