2,4-Dichloro-3-cyanophenylboronic acid 98%

Supplier: Ambeed
Warning

A803176-100MG A803176-250MG
77036-422EA 14.86 USD
77036-422 77036-424
2,4-Dichloro-3-cyanophenylboronic acid 98%
2,4-Dichloro-3-cyanophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₄BCl₂NO₂
MW: 215.83 g/mol
MDL Number: MFCD09800881
CAS Number: 957120-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR