Tetrahexylammonium bromide 98%

Supplier: Ambeed
Warning

A802934-1G A802934-5G A802934-25G A802934-100G A802934-10G
77036-420EA 11 USD
77036-420 77595-280 77603-036 77674-170 77674-172
Tetrahexylammonium bromide 98%
Tetrahexylammonium bromide

Precaución: for Research Use Only

Formula: C₂₄H₅₂BrN
MW: 434.59 g/mol
Melting Pt: 104
MDL Number: MFCD00011858
CAS Number: 4328-13-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR