4-Boc-1-(6-bromo-2-pyridyl)piperazine 97%

Supplier: Ambeed
Danger

A802662-1G A802662-250MG A802662-5G
76993-296EA 22 USD
76993-296 76993-298 76993-300
4-Boc-1-(6-bromo-2-pyridyl)piperazine 97%
4-Boc-1-(6-bromo-2-pyridyl)piperazine

Precaución: for Research Use Only

Formula: C₁₄H₂₀BrN₃O₂
MDL Number: MFCD07369773
CAS Number: 331767-56-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR