trans-Ethyl 3-aminocyclobutanecarboxylate hydrochloride 98%

Supplier: Ambeed
Warning

A802453-100MG A802453-250MG A802453-1G
76993-292EA 47 USD
76993-292 76993-294 77886-788
trans-Ethyl 3-aminocyclobutanecarboxylate hydrochloride 98%
trans-Ethyl 3-aminocyclobutanecarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₄ClNO₂
MDL Number: MFCD12925786
CAS Number: 957793-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR