3-Boc-aminomethylpyrrolidine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Boc-aminomethyl-pyrrolidine

A802153-100MG A802153-1G A802153-250MG A802153-5G A802153-10G
77019-422EA 22 USD
77019-422 77019-424 77019-426 77573-570 77891-552
3-Boc-aminomethylpyrrolidine 98%
3-Boc-aminomethylpyrrolidine

Precaución: for Research Use Only

Formula: C10H20N2O2
MW: 200.2780 g/mol
MDL Number: MFCD04973997
CAS Number: 149366-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR