Sodium-6,7-dihydroxynaphthalene-2-sulfonate 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydroxynaphthalene-6-sulphonic acid Monosodium Salt, DS acid Monosodium Salt, Sodium 2,3-dihydroxynaphthalene-6-sulphonate, Monosodium 6,7-Dihydroxynaphthalene-2-sulphonate

A802088-1000G A802088-100G A802088-25G A802088-500G
77006-580EA 199 USD
77006-580 77006-582 77006-584 77006-586
Sodium-6,7-dihydroxynaphthalene-2-sulfonate 98%
Sodium-6,7-dihydroxynaphthalene-2-sulfonate

Precaución: for Research Use Only

Formula: C₁₀H₈O₅S
MW: 262.21 g/mol
MDL Number: MFCD00004074
CAS Number: 135-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR