4-Hydroxy-5-nitronicotinic acid 97%

Supplier: Ambeed
Warning

A802014-100MG A802014-1G A802014-250MG
77036-408EA 78 USD
77036-408 77036-410 77036-412
4-Hydroxy-5-nitronicotinic acid 97%
4-Hydroxy-5-nitronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄N₂O₅
MDL Number: MFCD11044357
CAS Number: 911461-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR