7-Chloro-3,4-dihydroisoquinolin-1(2H)-one 96%
Supplier: Ambeed
|
Warning
|
A801452-100MG
A801452-1G
A801452-250MG
A801452-5G
A801452-10G
77019-414EA
8
USD
77019-414
77019-416
77019-418
77019-420
77891-551
7-Chloro-3,4-dihydroisoquinolin-1(2H)-one 96%
7-Chloro-3,4-dihydroisoquinolin-1(2H)-one
Precaución: for Research Use Only
Formula:
C₉H₈ClNO |
MDL Number:
MFCD04114870 CAS Number: 22245-95-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...