4-Chloro-6-(piperidin-1-yl)pyrimidin-5-amine 95%

Supplier: Ambeed
Warning

A801283-1G A801283-250MG
77006-570EA 264.17 USD
77006-570 77006-572
4-Chloro-6-(piperidin-1-yl)pyrimidin-5-amine 95%
4-Chloro-6-(piperidin-1-yl)pyrimidin-5-amine

Precaución: for Research Use Only

Formula: C₉H₁₃ClN₄
MW: 212.67 g/mol
MDL Number: MFCD04035010
CAS Number: 84762-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR