6-Morpholinobenzo[d]thiazol-2-amine 95%

Supplier: Ambeed
Warning

A801145-100MG A801145-1G A801145-250MG A801145-5G
77019-406EA 83.82 USD
77019-406 77019-408 77019-410 77019-412
6-Morpholinobenzo[d]thiazol-2-amine 95%
6-Morpholinobenzo[d]thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₃N₃OS
MW: 235.30 g/mol
MDL Number: MFCD03848248
CAS Number: 94641-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR