3-Amino-4-nitrophenol 95%

Supplier: Ambeed
Warning

A801031-100MG A801031-1G A801031-250MG A801031-5G
77036-394EA 13 USD
77036-394 77036-396 77036-398 77036-400
3-Amino-4-nitrophenol 95%
3-Amino-4-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
MDL Number: MFCD13185928
CAS Number: 16292-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR