Methyl 2-cyano-4-fluoro-6-methylbenzoate 95%

Supplier: Ambeed
Danger

A800145-100MG A800145-250MG
76993-284EA 58.35 USD
76993-284 76993-286
Methyl 2-cyano-4-fluoro-6-methylbenzoate 95%
Methyl 2-cyano-4-fluoro-6-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₈FNO₂
MW: 193.17 g/mol
MDL Number: MFCD22571667
CAS Number: 877151-43-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR