Nɑ-Carbobenzoxy-DL-asparagine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Nɑ-Cbz-DL-asparagine, Z-DL-Asn-OH, N-Benzyloxycarbonyl-DL-asparagine, Cbz-DL-Asn-OH, 4-Amino-2-(Cbz-amino)-4-oxobutyric Acid
A799871-100G
A799871-1G
A799871-25G
A799871-5G
77006-562EA
145
USD
77006-562
77006-564
77006-566
77006-568
Nɑ-Carbobenzoxy-DL-asparagine 98%
Nalpha-Carbobenzoxy-DL-asparagine
Precaución: for Research Use Only
Formula:
C₁₂H₁₄N₂O₅ MW: 266.25 g/mol |
MDL Number:
MFCD00065101 CAS Number: 29880-22-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...