1,2-Distearoyl-sn-glycero-3-phosphoethanolamine 95%

Supplier: Ambeed
Warning

Synonyms: 1,2-Distearoyl-sn-glycero-3-phosphoethanolamine, DSPE, 1,2-Dioctadecanoyl-sn-glycero-3-phosphoethanolamine, (2R)-3-(((2-Aminoethoxy)(hydroxy)phosphoryl)oxy)propane-1,2-diyl distearate, 2-Aminoethyl (R)-2,3-Bis(octadecanoyloxy)propyl Hydrogen Phosphate

A799800-10G A799800-1G A799800-5G A799800-100MG A799800-250MG
77019-396EA 1737.76 USD
77019-396 77019-398 77019-400 77614-426 77614-428
1,2-Distearoyl-sn-glycero-3-phosphoethanolamine 95%
1,2-Distearoyl-sn-glycero-3-phosphoethanolamine

Precaución: for Research Use Only

Formula: C₄₁H₈₂NO₈P
MW: 748.08 g/mol
MDL Number: MFCD00036777
CAS Number: 1069-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR