4-(tert-Butoxycarbonyl)-6-oxopiperazine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A799413-100MG A799413-1G A799413-250MG
76993-276EA 160.2 USD
76993-276 76993-278 76993-280
4-(tert-Butoxycarbonyl)-6-oxopiperazine-2-carboxylic acid 95%
4-(tert-Butoxycarbonyl)-6-oxopiperazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O₅
MW: 244.24 g/mol
MDL Number: MFCD10698236
CAS Number: 888220-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR