2-Chloro-3-fluoroisonicotinaldehyde 96%

Supplier: Ambeed
Warning

A798776-25G A798776-100G A798776-1G
77006-560EA 138 USD
77006-560 77882-720 77882-721
2-Chloro-3-fluoroisonicotinaldehyde 96%
2-Chloro-3-fluoroisonicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₃ClFNO
MDL Number: MFCD04972388
CAS Number: 329794-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR