Methyl-3-iodo-1H-indazole-4-carboxylate 95+%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-iodo-1H-indazole-4-carboxylate

A798518-100MG A798518-1G A798518-250MG A798518-5G
77036-372EA 18.04 USD
77036-372 77036-374 77036-376 77036-378
Methyl-3-iodo-1H-indazole-4-carboxylate 95+%
Methyl 3-iodo-1H-indazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇IN₂O₂
MW: 302.0686 g/mol
MDL Number: MFCD07781588
CAS Number: 885521-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR