7-Fluoro-3-iodo-1H-indazole 98%
Supplier: Ambeed
|
Warning
|
A798158-1G
A798158-250MG
A798158-5G
77036-366EA
20
USD
77036-366
77036-368
77036-370
7-Fluoro-3-iodo-1H-indazole 98%
7-Fluoro-3-iodo-1H-indazole
Precaución: for Research Use Only
Formula:
C₇H₄FIN₂ |
MDL Number:
MFCD10696806 CAS Number: 944904-38-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...