6-Bromo-2,3-dimethylpyridine 95%

Supplier: Ambeed
Warning

A797807-100MG A797807-250MG
76993-272EA 146.41 USD
76993-272 76993-274
6-Bromo-2,3-dimethylpyridine 95%
6-Bromo-2,3-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈BrN
MW: 186.04 g/mol
MDL Number: MFCD11869959
CAS Number: 99132-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR