2,2-Dimethylbenzo[d][1,3]dioxol-5-amine 95%

Supplier: Ambeed
Warning

A797322-1G A797322-250MG
76993-268EA 262.05 USD
76993-268 76993-270
2,2-Dimethylbenzo[d][1,3]dioxol-5-amine 95%
2,2-Dimethylbenzo[d][1,3]dioxol-5-amine

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.18 g/mol
MDL Number: MFCD03821918
CAS Number: 6324-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR