5-(2,5-Dimethyl-1H-pyrrol-1-yl)-2-hydroxybenzoic acid 95%

Supplier: Ambeed
Warning

A797247-100MG A797247-1G A797247-250MG
77006-552EA 56 USD
77006-552 77006-554 77006-556
5-(2,5-Dimethyl-1H-pyrrol-1-yl)-2-hydroxybenzoic acid 95%
5-(2,5-Dimethyl-1H-pyrrol-1-yl)-2-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₃
MDL Number: MFCD01125266
CAS Number: 313701-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR