2-(2-Bromo-4-fluorophenyl)-1,3-dioxolane 95+%

Supplier: Ambeed
Warning

A797058-1G A797058-250MG A797058-5G
77019-378EA 167.63 USD
77019-378 77019-380 77019-382
2-(2-Bromo-4-fluorophenyl)-1,3-dioxolane 95+%
2-(2-Bromo-4-fluorophenyl)-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MW: 247.06 g/mol
MDL Number: MFCD06209808
CAS Number: 773097-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR