2-Pyrrolidin-1-ylpropanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-(1-Pyrrolidyl)propanoic acid

A796711-1G A796711-5G
77036-358EA 80.63 USD
77036-358 77036-360
2-Pyrrolidin-1-ylpropanoic acid 95%
2-Pyrrolidin-1-ylpropanoic acid

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MW: 143.19 g/mol
MDL Number: MFCD05237232
CAS Number: 123912-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR