1-(2,4,6-Triisopropylphenylsulfonyl)-1,2,4-triazole 98%

Supplier: Ambeed
Warning

A796507-10G A796507-25G A796507-5G
77036-350EA 26 USD
77036-350 77036-352 77036-354 77036-356
1-(2,4,6-Triisopropylphenylsulfonyl)-1,2,4-triazole 98%
1-(2,4,6-Triisopropylphenylsulphonyl)-1,2,4-triazole

Precaución: for Research Use Only

Formula: C₁₇H₂₅N₃O₂S
MDL Number: MFCD00014089
CAS Number: 54230-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR