1-(6-Chloropyridin-3-yl)butan-1-one 95%
Supplier: Ambeed
|
Warning
|
A796134-100MG
A796134-1G
A796134-250MG
76993-258EA
30
USD
76993-258
76993-260
76993-262
1-(6-Chloropyridin-3-yl)butan-1-one 95%
1-(6-Chloropyridin-3-yl)butan-1-one
Precaución: for Research Use Only
Formula:
C₉H₁₀ClNO MW: 183.64 g/mol |
MDL Number:
MFCD13248626 CAS Number: 918503-72-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...