(3-Bromo-2-isopropoxy-5-methylphenyl)boronic acid 97%

Supplier: Ambeed
Warning

A795873-1G A795873-250MG A795873-5G
77019-372EA 65 USD
77019-372 77019-374 77019-376
(3-Bromo-2-isopropoxy-5-methylphenyl)boronic acid 97%
(3-Bromo-2-isopropoxy-5-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BBrO₃
MDL Number: MFCD04974111
CAS Number: 870718-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR