4-Hydroxy-3-pyridinesulfonic acid 97%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxypyridine-3-sulphonic acid

A795796-100G A795796-10G A795796-25G A795796-500G A795796-5G
77019-362EA 161.26 USD
77019-362 77019-364 77019-366 77019-368 77684-738
4-Hydroxy-3-pyridinesulfonic acid 97%, Ambeed.Inc
4-Hydroxy-3-pyridinesulphonic acid

Precaución: For research use only.

Formula: C₅H₅NO₄S
MW: 175.1625 g/mol
MDL Number: MFCD02093473
CAS Number: 51498-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR