1,1'-([1,1'-Biphenyl]-4,4'-diyl)bis(2-bromoethanone) 95%

Supplier: Ambeed
Danger

A795716-1G A795716-25G A795716-5G
77019-354EA 5 USD
77019-354 77019-356 77019-358
1,1'-([1,1'-Biphenyl]-4,4'-diyl)bis(2-bromoethanone) 95%
1,1'-([1,1'-Biphenyl]-4,4'-diyl)bis(2-bromoethanone)

Precaución: for Research Use Only

Formula: C₁₆H₁₂Br₂O₂
MDL Number: MFCD00017877
CAS Number: 4072-67-7
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR