4-Bromo-2-fluoro-5-nitrobenzaldehyde 95%

Supplier: Ambeed
Warning

A795656-1G A795656-250MG A795656-25G
77006-542EA 25 USD
77006-542 77006-544 77882-714
4-Bromo-2-fluoro-5-nitrobenzaldehyde 95%
4-Bromo-2-fluoro-5-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrFNO₃
MDL Number: MFCD10758067
CAS Number: 679839-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR