2-Chloro-6-(trifluoromethyl)nicotinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-6-(trifluoromethyl)pyridine-3-carboxylic acid

A795572-100MG A795572-10G A795572-1G A795572-250MG A795572-25G A795572-5G
77006-530EA 8.49 USD
77006-530 77006-532 77006-534 77006-536 77006-538 77006-540
2-Chloro-6-(trifluoromethyl)nicotinic acid 97%
2-Chloro-6-(trifluoromethyl)nicotinic acid

Precaución: for Research Use Only

Formula: C₇H₃ClF₃NO₂
MW: 225.56 g/mol
MDL Number: MFCD01862658
CAS Number: 280566-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR