AKR1C3 Inhibitor I 98%

Supplier: Ambeed
Warning

A795496-100MG A795496-1G A795496-250MG
76993-252EA 33 USD
76993-252 76993-254 76993-256
AKR1C3 Inhibitor I 98%
AKR1C3 Inhibitor I

Precaución: for Research Use Only

Formula: C₁₆H₁₅NO₄S
MDL Number: MFCD01197550
CAS Number: 327092-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR