1-Cyclopropyl-6-fluoro-8-methoxy-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
PD 135042
A795037-100MG
A795037-250MG
76993-248EA
76.39
USD
76993-248
76993-250
1-Cyclopropyl-6-fluoro-8-methoxy-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid 95%
1-Cyclopropyl-6-fluoro-8-methoxy-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₈H₂₀FN₃O₄ MW: 361.37 g/mol |
MDL Number:
MFCD25973505 CAS Number: 112811-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...