3-Fluoro-2-(trifluoromethyl)pyridine 97%

Supplier: Ambeed
Danger

A793898-1G A793898-5G
77036-340EA 9 USD
77036-340 77873-258
3-Fluoro-2-(trifluoromethyl)pyridine 97%
3-Fluoro-2-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₃F₄N
MDL Number: MFCD08437598
CAS Number: 886510-21-0
UN: 1992
ADR: 3,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR