tert-Butyl 3,5-dimethyl-4-oxopiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A793529-100MG A793529-5G
76993-246EA 116.7 USD
76993-246 77573-540
tert-Butyl 3,5-dimethyl-4-oxopiperidine-1-carboxylate 95%
tert-Butyl 3,5-dimethyl-4-oxopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₃
MW: 227.30 g/mol
MDL Number: MFCD14635714
CAS Number: 475085-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR