5-Chloro-6-(trifluoromethyl)-1H-indole 95%

Supplier: Ambeed
Warning

A792715-100MG A792715-1G A792715-250MG
77006-510EA 92.3 USD
77006-510 77006-512 77006-514
5-Chloro-6-(trifluoromethyl)-1H-indole 95%
5-Chloro-6-(trifluoromethyl)-1H-indole

Precaución: for Research Use Only

Formula: C₉H₅ClF₃N
MDL Number: MFCD03094972
CAS Number: 1186404-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR