1,8-Bis(diphenylphosphino)octane 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A792621-250MG A792621-1G A792621-5G
76993-244EA 34 USD
76993-244 77573-534 77684-735
1,8-Bis(diphenylphosphino)octane 98%, Ambeed.Inc
1,8-Bis(diphenylphosphino)octane

Precaución: For research use only.

Formula: C₃₂H₃₆P₂
MDL Number: MFCD09834841
CAS Number: 41625-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR