5-Amino-6-bromoquinoline 98%

Supplier: Ambeed
Danger

Synonyms: 6-Bromoquinolin-5-amine

A792500-100MG A792500-250MG A792500-1G A792500-5G
77036-336EA 37 USD
77036-336 77036-338 77873-251 77873-252
5-Amino-6-bromoquinoline 98%
5-Amino-6-bromoquinoline

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MDL Number: MFCD11110424
CAS Number: 50358-39-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR