3-(Trifluoromethyl)-1H-1,2,4-triazole 95%
Supplier: Ambeed
|
Warning
|
A792176-100MG
A792176-250MG
A792176-1G
A792176-5G
77006-502EA
20
USD
77006-502
77006-504
77595-258
77595-260
3-(Trifluoromethyl)-1H-1,2,4-triazole 95%
3-(Trifluoromethyl)-1H-1,2,4-triazole
Precaución: for Research Use Only
Formula:
C₃H₂F₃N₃ |
MDL Number:
MFCD11847494 CAS Number: 60406-75-9 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...