2-Chloro-4-iodobenzonitrile 95%

Supplier: Ambeed
Warning

A791660-100MG A791660-1G A791660-250MG
76993-234EA 42.44 USD
76993-234 76993-236 76993-238
2-Chloro-4-iodobenzonitrile 95%
2-Chloro-4-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃ClIN
MW: 263.46 g/mol
MDL Number: MFCD18393346
CAS Number: 371764-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR