Dibenzosuberenone 98%

Supplier: Ambeed
Warning

Synonyms: 5H-Dibenzo[a,d]cyclohepten-5-one, 5H-Dibenzo[a,d][7]annulen-5-one

A791387-100G A791387-10G A791387-25G A791387-500G A791387-5G
77006-486EA 59.42 USD
77006-486 77006-488 77006-490 77006-492 77006-494
Dibenzosuberenone 98%
Dibenzosuberenone

Precaución: for Research Use Only

Formula: C₁₅H₁₀O
MW: 206.24 g/mol
MDL Number: MFCD00003588
CAS Number: 2222-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR