2-Chloro-N-(2-chloro-4-methyl-3-pyridyl)nicotinamide 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-N-(2-chloro-4-methylpyridin-3-yl)pyridine-3-carboxamide, 2-Chloro-N-(2-chloro-4-methylpyridin-3-yl)nicotinamide

A791075-1G A791075-25G A791075-5G
77006-480EA 24.41 USD
77006-480 77006-482 77006-484
2-Chloro-N-(2-chloro-4-methyl-3-pyridyl)nicotinamide 98%
2-Chloro-N-(2-chloro-4-methyl-3-pyridyl)nicotinamide

Precaución: for Research Use Only

Formula: C₁₂H₉Cl₂N₃O
MW: 282.12 g/mol
MDL Number: MFCD09031256
CAS Number: 133627-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR