2-(2,3-Dimethylphenoxy)acetohydrazide 95%

Supplier: Ambeed
Warning

A790974-1G A790974-250MG A790974-5G
77036-330EA 118.83 USD
77036-330 77036-332 77036-334
2-(2,3-Dimethylphenoxy)acetohydrazide 95%
2-(2,3-Dimethylphenoxy)acetohydrazide

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₂
MDL Number: MFCD01536449
CAS Number: 134432-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR