4-Aminopyrrolidin-2-one hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A790944-100MG
A790944-1G
A790944-250MG
A790944-5G
77036-322EA
12
USD
77036-322
77036-324
77036-326
77036-328
4-Aminopyrrolidin-2-one hydrochloride 97%
4-Aminopyrrolidin-2-one hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₉ClN₂O |
MDL Number:
MFCD11519397 CAS Number: 167465-93-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...