4-Acridinecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Acridine-4-carboxylic acid

A790146-100MG A790146-1G A790146-250MG
76993-228EA 15 USD
76993-228 76993-230 76993-232
4-Acridinecarboxylic acid 98%
4-Acridinecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₉NO₂
MW: 223.227 g/mol
MDL Number: MFCD21340176
CAS Number: 31327-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR