2-Bromo-N-phenyl-acetamide 97%

Supplier: Ambeed
Danger

A789931-100MG A789931-250MG A789931-1G
77036-318EA 14 USD
77036-318 77036-320 77573-528
2-Bromo-N-phenyl-acetamide 97%
2-Bromo-N-phenyl-acetamide

Precaución: for Research Use Only

Formula: C₈H₈BrNO
MDL Number: MFCD00462518
CAS Number: 5326-87-4
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR