2-((Dimethylamino)methyl)acrylic acid 95%

Supplier: Ambeed
Warning

A789686-100MG A789686-1G A789686-250MG
76993-218EA 129 USD
76993-218 76993-220 76993-222
2-((Dimethylamino)methyl)acrylic acid 95%
2-((Dimethylamino)methyl)acrylic acid

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
MDL Number: MFCD19656217
CAS Number: 5415-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR