N-Cyclohexyl 3-boronobenzenesulfonamide 97%

Supplier: Ambeed
Warning

Synonyms: (3-(N-Cyclohexylsulphamoyl)phenyl)boronic acid

A788942-100MG A788942-1G A788942-250MG
77019-316EA 36.08 USD
77019-316 77019-318 77019-320
N-Cyclohexyl 3-boronobenzenesulfonamide 97%
N-Cyclohexyl 3-boronobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₁₂H₁₈BNO₄S
MW: 283.1516 g/mol
MDL Number: MFCD07783859
CAS Number: 871329-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR